

1-Propoxy-2-propanol ≥93.0% (by GC) (contains 2-Isopropoxy-1-propanol)
Supplier: TCI
Certificates
About this item
CAS Number:1569-01-3
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00192420
:118.18 g/mol
Boiling Pt:149°C
Flash Pt:48°C
EINECS:216-372-4
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 93.0 %
Specification Test Results
Frequently Bought Together