Order Entry
Netherlands
ContactUsLinkComponent
4-Propylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
 
 
4-Propylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
  TCIAP2022-5ML
 :  TCI
 :  
undefined
 
 
4-Propylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
  TCIAP2022-5ML
 :  TCI
 :  

 

  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :141.26 g/mol
 :2733
 :3,III
 :GHS02
 :GHS05
 :102653-37-2
 :C₉H₁₉N
 :MFCD11156904