[6,6]-Phenyl-C61-butyric acid dodecyl ester ≥98.0% (by HPLC)
Catalog # TCIAP2015-100MG
Supplier: TCI
CAS Number:
Specifications
- VPE:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:1065.20 g/mol
CAS Number:571177-69-0
Formula:C₈₃H₃₆O₂