[6,6]-Phenyl-C61-butyric acid n-octyl ester ≥98.0% (by HPLC)
Catalog # TCIAP2014-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:1009.09 g/mol
CAS Number:571177-68-9
Formula:C₇₉H₂₈O₂
MDL Number:MFCD09265175