
3-(3-Pyridyl)propionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3724-19-4
:
Formula:C₈H₉NO₂
MDL Number:MFCD00274197
:151.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:156.0 to 160.0 deg-C
Specification Test Results
Frequently Bought Together