Order Entry
Netherlands
ContactUsLinkComponent
1,10-Phenanthroline-5,6-dione ≥98.0% (by HPLC, titration analysis)
Irritant
1,10-Phenanthroline-5,6-dione ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1,10-Phenanthroline-5,6-dione ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together