Order Entry
Netherlands
ContactUsLinkComponent
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride ≥98.0% (by HPLC, total nitrogen)
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    189.0 to 195.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together