Order Entry
Netherlands
ContactUsLinkComponent
Propyne ∼3-4% in heptane
Flammable
Irritant
Health
Propyne ∼3-4% in heptane
Catalog # TCIAP1881-200G
Supplier:  TCI
CAS Number:  
Flammable
Irritant
Health
Propyne ∼3-4% in heptane
Catalog # TCIAP1881-200G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 g
  • Seq:
    3505

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (GC):
    3.0 to 4.0 w/w%

Specification Test Results

About this item

 :
 :40.06 g/mol
Storage Temperature:Freezer
EINECS:200-828-4
UN:1206
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:74-99-7
Formula:C₃H₄
MDL Number:MFCD00036235