
Phenyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17763-67-6
:
Formula:C₇H₅F₃O₃S
MDL Number:MFCD00192399
:226.17 g/mol
Boiling Pt:62°C
Flash Pt:71°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together