Order Entry
Netherlands
ContactUsLinkComponent
L-Proline-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Irritant
L-Proline-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI
Certificates
Irritant
L-Proline-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    109.0 to 114.0 deg-C
  • Specific rotation [a]20/D:
    -30.0 to -33.0 deg (C=1、H2O)

Specification Test Results

Related Information

Frequently Bought Together