γ-(N-Morpholino)propanesulphonic acid (MOPS) 99.5% for molecular biology, for peptide sequencing DNase-, RNase-, protease-free
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1132-61-2
:
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
:209.27 g/mol
Merck Index:13,06288
Melting Pt:277…282 °C
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:214-478-5
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Assay:≥99.5 %
- Loss on drying:≤0.5 % (105°C)
- Solubility:(10 % in water) Complete
- UV:(0.1 M in water)
- pH:3.0 to 4.5 (1 % at 25°C)
- Dnase,Rnase,Protease activity:none detected
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together