Phytyl acetate (cis and trans mixture) cis- and trans-mixture ≥88.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 3,7,11,15-Tetramethyl-2-hexadecenyl Acetate, Acetic Acid 3,7,11,15-Tetramethyl-2-hexadecenyl Ester
CAS Number:10236-16-5
:
Formula:C₂₂H₄₂O₂
Molecular Weight:338.58 g/mol
Flash Pt:184°C
EINECS:233-565-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 88.0 % (total of isomers)
Specification Test Results
Frequently Bought Together