
2-(4-Pyridyl)benzimidazole ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:2208-59-5
:
Formula:C₁₂H₉N₃
MDL Number:MFCD00453948
:195.23 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:216.0 to 220.0 deg-C
Specification Test Results
Frequently Bought Together