Order Entry
Netherlands
ContactUsLinkComponent
5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione ≥93.0% (by HPLC)
5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione ≥93.0% (by HPLC)
Supplier:  TCI
Certificates
5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione ≥93.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 93.0 area%
  • Melting point:
    49.0 to 53.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together