
Pyridinium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52193-54-1
:
Formula:C₆H₆F₃NO₃S
MDL Number:MFCD00274656
:229.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:222.0 to 226.0 deg-C
Specification Test Results
Frequently Bought Together