
Pentafluoroanilinium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:912823-79-1
:
Formula:C₇H₃F₈NO₃S
MDL Number:MFCD08276416
:333.15 g/mol
UN:1759
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together