

1,3-Propanediamine-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
: TCI
:1939-36-2
:
:C₁₁H₁₈N₂O₈
:MFCD00210039
:306.27 g/mol
:400-400-9
:GHS05
:GHS07
- Purity (Chelometric Titration):min. 98.0 %
- Appearance:White to Almost white powder to crystal
- Solubility in 1mol/L NaOH:almost transparency
Frequently Bought Together