
2-(4-Pyridyl)-2-propanol ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15031-78-4
:
Formula:C₈H₁₁NO
MDL Number:MFCD00071746
:137.18 g/mol
Boiling Pt:140°C
Melting Pt:136°C
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:133.0 to 138.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together