
2-(4-Piperidyl)-2-propanol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:22990-34-7
:
Formula:C₈H₁₇NO
MDL Number:MFCD06797159
:143.23 g/mol
Melting Pt:136°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
Specification Test Results
Frequently Bought Together