4-Propargylthiomorpholine-1,1-dioxide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10442-03-2
:
Formula:C₇H₁₁NO₂S
MDL Number:MFCD00085157
:173.23 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together