1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)dibromide ≥98.0% (by HPLC)
: TCI
:106867-97-4
:
:C₂₈H₂₄Br₂N₄
:MFCD03844804
:576.34 g/mol
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Frequently Bought Together