(1S,2R)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenylpropyl propionate ≥98.0% (by HPLC) reagent for anti-selective asymmetric aldol reaction
Catalog # TCIAP1372-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:147.0 to 150.0 deg-C
- Specific rotation [a]20/D:-8.0 to -11.0 deg (C=2, Chloroform)
Specification Test Results
About this item
:
:479.64 g/mol
CAS Number:187324-67-0
Formula:C₂₈H₃₃NO₄S
MDL Number:MFCD02093491