(1R,2S)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenylpropyl propionate ≥98.0% (by HPLC) reagent for anti-selective asymmetric aldol reaction
TCIAP1371-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:147.0 to 150.0 deg-C
- Specific rotation [a]20/D:+8.0 to +11.0 deg (C=2, CHCl3)
:
:479.64 g/mol
:187324-66-9
:C₂₈H₃₃NO₄S
:MFCD02093490