
Pyridine-2,3-dicarboxamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:4663-94-9
:
Formula:C₇H₇N₃O₂
MDL Number:MFCD00191488
:165.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together