Order Entry
Netherlands
ContactUsLinkComponent
 
Toxic
Health
Environment
o-Phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
Catalog # TCIAP1144-1G
Supplier:  TCI
CAS Number:  
undefined
Toxic
Health
Environment
o-Phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
Catalog # TCIAP1144-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Absorbance:
    max. 0.040 (1w/v%, H2O, 420.0 nm)
  • Absorbance2:
    max. 0.050 (1w/v%, H2O, 480.0 nm)

Specification Test Results

About this item

 :
 :181.06 g/mol
Melting Pt:205°C
EINECS:210-418-7
UN:2811
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:615-28-1
Formula:C₆H₁₀Cl₂N₂
MDL Number:MFCD00012966