Order Entry
Netherlands
ContactUsLinkComponent
trans-3-Pentenoic acid ≥95.0% (by GC, titration analysis)
Corrosive
trans-3-Pentenoic acid ≥95.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Corrosive
trans-3-Pentenoic acid ≥95.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together