
(±)-Propranolol hydrochloride ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:318-98-9
:3506-09-0
Formula:C₁₆H₂₁NO₂·HCl
MDL Number:MFCD00012558
:295.81 g/mol
Merck Index:14,07840
EINECS:206-268-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:163.0 to 168.0 deg-C
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together