cis-3-Hexenyl pyruvate ≥95.0% (by GC, titration analysis)
Catalog # TCIAP0894-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
- Purity (Iodometric Titration):min. 95.0 %
Specification Test Results
About this item
:
:170.21 g/mol
Boiling Pt:98°C
Flash Pt:108°C
Storage Temperature:Refrigerator
EINECS:268-703-7
CAS Number:68133-76-6
Formula:C₉H₁₄O₃
MDL Number:MFCD00036527