
N,N'-1,4-Phenylenedimaleimide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3278-31-7
:
Formula:C₁₄H₈N₂O₄
MDL Number:MFCD00005505
:268.23 g/mol
EINECS:221-910-6
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot NaOH:almost transparency
Specification Test Results
Frequently Bought Together