
Polyethylene glycol monooleyl ether n=∼2
Supplier: TCI
Certificates
About this item
CAS Number:9004-98-2
:
Formula:HO(CH₂CH₂O)nC₁₈H₃₅
MDL Number:MFCD00074369
EINECS:500-016-2
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- The average value of n:1.8 to 2.2
Specification Test Results
Frequently Bought Together