
Polyethylene glycol mono-4-nonylphenyl ether n=∼18
Catalog # TCIAP0709-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white wax
- Acid Value:max. 1.0
- The average value of n:17.000 to 20.000
Specification Test Results
About this item
:
Merck Index:14,06677
EINECS:500-045-0
:GHS07
CAS Number:26027-38-3
MDL Number:MFCD00148006