
1-(4-Pyridyl)pyridinium chloride hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5421-92-1
:
Formula:C₁₀H₉ClN₂·HCl·xH₂O
MDL Number:MFCD00012821
:229.10 g/mol (anhy)
EINECS:226-543-5
:GHS07
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:3.0 to 10.0 %
Specification Test Results
Frequently Bought Together