Piperazine citrate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:144-29-6
:
Formula:C₄H₁₀N₂
MDL Number:MFCD00038961
:642.66 g/mol
Merck Index:14,07464
EINECS:205-622-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 12.0 %
Specification Test Results
Frequently Bought Together