Phenylthiohydantoin-δ-threonine ≥90.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:5800-50-0
:
Formula:C₁₁H₁₀N₂OS
MDL Number:MFCD00070617
:218.27 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 90.0 area%
- Melting point:234.0 to 238.0 deg-C
Specification Test Results
Frequently Bought Together