
Phosphocholine chloride calcium salt tetrahydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:72556-74-2
:MFCD00149983
Formula:C₅H₁₃CaClNO₄P·4H₂O
:257.67 g/mol (anhy)
Merck Index:14,07362
EINECS:225-403-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 % (calcd.on anh.substance)
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh.substance)
- Water:19.0 to 24.0 %
Specification Test Results
Frequently Bought Together