


Tris(1,3-dichloro-2-propyl)phosphate ≥93.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13674-87-8
:
Formula:C₉H₁₅Cl₆O₄P
MDL Number:MFCD00083121
:430.89 g/mol
Merck Index:14,09087
Boiling Pt:252°C
Flash Pt:249°C
EINECS:237-159-2
UN:3082
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 93.0 %
Specification Test Results
Frequently Bought Together