


Dibutyl phosphate (mono- and di-ester mixture) (Mono- and Di- Ester mixture)
Supplier: TCI
Certificates
About this item
CAS Number:107-66-4
:
Formula:C₈H₁₉O₄P
MDL Number:MFCD00015245
:210.21 g/mol
Flash Pt:180°C
EINECS:203-509-8
UN:1718
ADR:8,III
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Assay of mono ester:34.0 to 45.0 %
- Assay of Diester:55.0 to 66.0 %
Specification Test Results
Frequently Bought Together