Order Entry
Netherlands
ContactUsLinkComponent
Methyl phosphate (mono and di ester mixture)
Corrosive
Methyl phosphate (mono and di ester mixture)
Catalog # TCIAP0262-25ML
Supplier:  TCI
CAS Number:  
Corrosive
Methyl phosphate (mono and di ester mixture)
Catalog # TCIAP0262-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Assay of mono ester:
    50.0 to 55.0 %
  • Assay of Diester:
    40.0 to 45.0 %

Specification Test Results

About this item

 :
Flash Pt:177°C
UN:3265
ADR:8,III
 :GHS05
CAS Number:52932-95-3
MDL Number:MFCD00015936