Phenylphosphonic acid disodium salt hydrate ≥98.0% (by titrimetric analysis)
TCIAP0205-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:very faint turbidity
- Water:max. 5.0 %
:
:202.06 g/mol
:25148-85-0
:C₆H₇O₃P
:MFCD00058972