



Phenylmagnesium Iodide ∼42% ∼2 mol/L in ethyl ether
Catalog # TCIAP0191-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):41.0 to 46.5 w/w%
Specification Test Results
About this item
:
:228.32 g/mol
UN:2924
ADR:3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:16002-63-4
Formula:C₆H₅IMg
MDL Number:MFCD00059490