Order Entry
Netherlands
ContactUsLinkComponent
 
2-Phenylazo-4-methylphenol ≥98.0% (by HPLC, total nitrogen)
  TCIAP0145-25G
 :  TCI
 :  
2-Phenylazo-4-methylphenol ≥98.0% (by HPLC, total nitrogen)
  TCIAP0145-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Orange to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    105.0 to 109.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

 

 

 :
 :212.25 g/mol
 :952-47-6
 :C₁₃H₁₂N₂O
 :MFCD00045788