[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:70331-94-1
:
Formula:C₄₀H₆₀N₂O₈
MDL Number:MFCD01940880
:696.93 g/mol
EINECS:274-572-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:178.0 to 182.0 deg-C
Specification Test Results
Frequently Bought Together