Order Entry
Netherlands
ContactUsLinkComponent
Sodium thiosulphate pentahydrate ≥99.0% (by titrimetric analysis)
Sodium thiosulphate pentahydrate ≥99.0% (by titrimetric analysis)
  TCIAO0522-300G
 :  TCI
 :  
Sodium thiosulphate pentahydrate ≥99.0% (by titrimetric analysis)
  TCIAO0522-300G
 :  TCI
 :  

 

  • Pk:
    300 g
  • Seq:
    3575

 

 

  • Appearance:
    White powder to crystal
  • Purity (Iodometric Titration):
    min. 99.0 %

 

 

 :
 :248.19 g/mol
 :48°C
 :10102-17-7
 :Na₂S₂O₃.5H₂O
 :MFCD00149186