
1H,1H,5H-Octafluoropentyl methacrylate ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:355-93-1
:
Formula:C₉H₈F₈O₂
MDL Number:MFCD00039278
:300.15 g/mol
Boiling Pt:88°C
Flash Pt:81°C
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:206-596-0
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together