
cis-Octahydroisoindole ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1470-99-1
:
Formula:C₈H₁₅N
MDL Number:MFCD07778620
:125.22 g/mol
Boiling Pt:189°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together