Order Entry
Netherlands
ContactUsLinkComponent
2-Oxa-6-azaspiro[3.3]heptane ≥98.0% (by GC)
Flammable
Irritant
2-Oxa-6-azaspiro[3.3]heptane ≥98.0% (by GC)
Catalog # TCIAO0466-1G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2-Oxa-6-azaspiro[3.3]heptane ≥98.0% (by GC)
Catalog # TCIAO0466-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :99.13 g/mol
Storage Temperature:Freezer
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:174-78-7
Formula:C₅H₉NO
MDL Number:MFCD12407093