
4-(Octyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAO0462-1G
Leverancier: TCI
CAS-nummer:
Specificaties
- Pk:1 g
- Seq:3047
Specificaties
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 107.0 %
Specificatie testresultaten
Over dit artikel
:
:250.1400 g/mol
Smelten Pt:73°C
:GHS07
CAS-nummer:121554-09-4
Formule:C₁₄H₂₃BO₃
MDL-nummer:MFCD00982004