(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate ≥98.0% (by GC)
Catalog # TCIAO0460-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:47.0 to 51.0 deg-C
- Specific rotation [a]20/D:-25.0 to -29.0 deg (C=5, THF)
Specification Test Results
About this item
:
:454.61 g/mol
CAS Number:133676-09-2
Formula:C₂₈H₃₈O₅
MDL Number:MFCD21603881