Order Entry
Netherlands
ContactUsLinkComponent
9-n-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
9-n-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAO0439-200MG
Supplier:  TCI
CAS Number:  
undefined
9-n-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAO0439-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

About this item

 :
 :531.35 g/mol
CAS Number:406726-92-9
Formula:C₃₂H₄₇B₂NO₄
MDL Number:MFCD23703114