
1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26482-00-8
:
Formula:C₁₂H₁₁IN₂O
MDL Number:MFCD01690884
:326.14 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together